Description
硫酸钠、硫酸铵及硫酸镍等硫酸盐,广泛应用于化学化工领域,本文采用了新型离子色谱连接方式-循环离子色谱概念,改进了以前柱切换需要两个泵和两个阀的管路连接方式,只需要单泵就能实现高浓度样品中痕量杂质离子的测定。采用KOH溶剂发生器能够较方便准确得到所需的淋洗液浓度,同时经过第一次预分离后,溶液通过抑制器已经转换成水溶液,从而保证氯离子在富集柱上有较强的保留,而不会有损失。该方法简单方便,可用于分析纯硫酸盐试剂中痕量氯离子的检验,对硫酸盐试剂进行质量控制,从而提高分析纯硫酸盐的产品质量。| Market: | Chemical |
| Keywords: | 离子色谱;阀切换硫酸盐(AR);氯离子 |
| Matrix: | 纯硫 酸盐固体 |
| Author: | 张婷婷 |
For Research Use Only. Not for use in diagnostic procedures.