Find methods for your needs
Meeting the Upcoming European Commission Performance Criteria for the Use of Triple Quadrupole GC-MS/MS as a Confirmatory Method for PCDD/Fs in Food and Feed Samples
# | Compound Name | Compound Class | RT (min) |
---|---|---|---|
1 | 2378-TCDF | tetra chlorinated dibenzo-furan | 16.56 |
2 | 2378-TCDD | tetra chlorinated dibenzo-p-dioxin | 16.92 |
3 | 12378-PeCDF | penta chlorinated dibenzo-furan | 19.18 |
4 | 23478-PeCDF | penta chlorinated dibenzo-furan | 19.89 |
5 | 12378-PeCDD | penta chlorinated dibenzo-p-dioxin | 20.13 |
6 | 123478-HxCDF | hexa chlorinated dibenzo-furan | 22.70 |
7 | 123678-HxCDF | hexa chlorinated dibenzo-furan | 22.83 |
8 | 234678-HxCDF | hexa chlorinated dibenzo-furan | 23.42 |
9 | 123478-HxCDD | hexa chlorinated dibenzo-p-dioxin | 23.69 |
10 | 123678-HxCDD | hexa chlorinated dibenzo-p-dioxin | 23.69 |
11 | 123789-HxCDD | hexa chlorinated dibenzo-p-dioxin | 24.02 |
12 | 123789-HxCDF | hexa chlorinated dibenzo-furan | 24.45 |
13 | 1234678-HpCDF | hepta chlorinated dibenzo-furan | 26.02 |
14 | 1234678-HpCDD | hepta chlorinated dibenzo-p-dioxin | 27.02 |
15 | 1234789-HpCDF | hepta chlorinated dibenzo-furan | 27.51 |
16 | OCDD | octa chlorinated dibenzo-p-dioxin | 29.51 |
17 | OCDF | octa chlorinated dibenzo-furans | 29.67 |
Description
Polychlorinated dibenzo-p-dioxins (PCDDs) and polychlorinated dibenzofurans (PCDFs) constitute a group of highly toxic organic compounds. PCDD/Fs have been characterized by the US Environmental Protection Agency as likely to be carcinogenic to humans. Recent technological advances in GC-MS/MS technology led to GC-MS/MS being considered a reliable tool that can be used to control the maximum levels for PCDD/Fs in food and feed. In this work, the performance of the Thermo Scientific TSQ 8000 Evo triple quadrupole GC-MS/MS for the analysis of PCDD/Fs was assessed.Market: | Food and Beverage |
Keywords: | GC-MS/MS, Quantification, Dioxins, food and feed, isotope dilution, TargetQuan |
Matrix: | Food |
For Research Use Only. Not for use in diagnostic procedures.