Find methods for your needs
TN27: Determination of Lanthanide Metals in Digested Rock Samples by Chelation Ion Chromatography
Description
A technique called chelation ion chromatography (chelation IC) was developed for the determination of transition and lanthanide metals in complex matrices. The sample pretreatment system (chelation concentration) coupled directly to the ion chromatograph offers the analyst a solution to the detection limit and interference problems commonly experienced when analyzing complex matrices.| Market: | GeoSciences |
| Keywords: | Chelation Ion Chromatography, Lanthanide Metals, IonPac CS5, Rock |
| Matrix: | Digested acid solution |
| Affiliation: | Thermo Scientific |
| Status: | Proof of Concept |
For Research Use Only. Not for use in diagnostic procedures.