Find methods for your needs
USP 38 monograph: impurity analysis of tranexamic acid using a C18 HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the impurity analysis of tranexamic acid. The analysis was performed on a Thermo Scientific Acclaim 120 C18 HPLC column using the method described in the USP 38 monograph. Resolution between tranexamic acid and impurity C was 3.9 (exceeding USP criteria of not less than 2.0).| Market: | Pharma |
| Keywords: | HPLC, Tranexamic acid, UV detection, USP 38 monograph |
| Matrix: | N/A |
| Author: | Manish Singh |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.