Find methods for your needs
Three-Fold Increase in Productivity for Pesticide Residue Analysis in Baby Food Using Fast Triple Quadrupole GC-MS/MS Be the first to rate this application
threefold increase productivity for pesticide residue analysis baby food using fast triple quadrupole gcmsms
# Compound Name Compound Class RT (min)
1 Dichlorvos Pesticide 4.33
2 Dichlobenil Pesticide 4.69
3 EPTC Pesticide 4.73
4 Biphenyl Pesticide 4.85
5 Etridiazole Pesticide 5.07
6 Methacrifos Pesticide 5.21
7 Diphenylamine Pesticide 5.68
8 Ethoprop (Ethoprophos) Pesticide 5.70
9 Cycloate Pesticide 5.72
10 Chlorpropham Pesticide 5.77
11 Dicrotophos Pesticide 5.79
12 Cadusafos Pesticide 5.88
13 BHC, Alpha Pesticide 5.96
14 Hexachlorobenzene Pesticide 6.00
15 Dichloran Pesticide 6.04
16 Chlorbufam Pesticide 6.09
17 BHC, Beta Pesticide 6.11
18 Clomazone Pesticide 6.12
19 BHC, gamma Pesticide 6.18
20 Diazinon Pesticide 6.21
21 Chlorothalonil Pesticide 6.29
22 Paraoxon-methyl Pesticide 6.30
23 Etrimfos Pesticide 6.33
24 Pentachloroaniline Pesticide 6.48
25 Dimethenamid Pesticide 6.52
26 Acetochlor Pesticide 6.53
27 Metribuzin Pesticide 6.54
28 Chlorpyrifos-methyl Pesticide 6.55
29 Malaoxon Pesticide 6.55
30 Parathion-methyl Pesticide 6.59
31 Heptachlor Pesticide 6.67
32 Ethofumesate Pesticide 6.74
33 Fenitrothion Pesticide 6.74
34 Pentanochlor (Solan) Pesticide 6.78
35 Dichlofluanid Pesticide 6.80
36 Chlorpyrifos-ethyl Pesticide 6.85
37 Cyanazine Pesticide 6.85
38 Parathion (ethyl) Pesticide 6.88
39 Aldrin Pesticide 6.90
40 Carbetamide Pesticide 6.90
41 Flurochloridone Pesticide 6.92
42 Nitrothal-isopropyl Pesticide 6.92
43 Pendimethalin Pesticide 7.04
44 Metazachlor Pesticide 7.06
45 Chlozolinate Pesticide 7.07
46 Mephosfolan Pesticide 7.11
47 Heptachlor epoxide-cis Pesticide 7.12
48 Oxychlordane Pesticide 7.12
49 Furalaxyl Pesticide 7.13
50 Heptachlor epoxide-trans Pesticide 7.14
51 Captan Pesticide 7.16
52 Folpet Pesticide 7.19
53 Bromophos-ethyl Pesticide 7.21
54 Methidathion Pesticide 7.21
55 Chlordane alpha-cis Pesticide 7.25
56 Chlordane gamma-trans Pesticide 7.32
57 Flutolanil Pesticide 7.33
58 Napropamide Pesticide 7.34
59 Endosulfan I Pesticide 7.36
60 Oxadiazon Pesticide 7.39
61 DDE p, p Pesticide 7.42
62 Oxyfluorfen Pesticide 7.42
63 Bupirimate Pesticide 7.43
64 Buprofezin Pesticide 7.45
65 Dieldrin Pesticide 7.47
66 Nitrofen Pesticide 7.54
67 Endrin Pesticide 7.58
68 Ethion Pesticide 7.61
69 Endosulfan II Pesticide 7.62
70 DDD p,p Pesticide 7.63
71 Aclonifen Pesticide 7.64
72 DDT o,p Pesticide 7.65
73 Benalaxyl Pesticide 7.74
74 DDT p,p Pesticide 7.80
75 Endosulfan sulfate Pesticide 7.81
76 Hexazinone Pesticide 7.83
77 Diflufenican Pesticide 7.86
78 Iprodione Pesticide 7.99
79 Bifenthrin Pesticide 8.00
80 EPN Pesticide 8.03
81 Bromopropylate Pesticide 8.04
82 Etoxazole Pesticide 8.04
83 Fenpropathrin Pesticide 8.05
84 Methoxychlor Pesticide 8.05
85 Bifenox Pesticide 8.09
86 Fenazaquin Pesticide 8.11
87 Phosalone Pesticide 8.18
88 Cyhalothrin-S Pesticide 8.23
89 Azinphos-ethyl Pesticide 8.34
90 Permethrin I Pesticide 8.43
91 Permethrin II Pesticide 8.46
92 Coumaphos Pesticide 8.48
93 Cyfluthrin peaks I-IV Pesticide 8.59
94 Cypermethrin peaks I-IV Pesticide 8.68
95 Flucythrinate I Pesticide 8.69
96 Flucythrinate II Pesticide 8.74
97 Fenvalerate I Pesticide 8.91
98 Fluvalinate Pesticide 8.93
99 Fenvalerate II Pesticide 8.98
100 Deltamethrin Pesticide 9.15
Description
In this study, a fast, easy, and robust workflow was used to analyze pesticide residues in baby food. Accurate and sensitive detection, quantification, and identification of pesticides in baby foods is of particular importance because babies are more vulnerable to adverse health effects from these chemicals.
Market: Food and Beverage
Keywords: Baby Food, Food Safety, GC-MS/MS, Pesticide Analysis, TraceFinder
Matrix: Baby food
Author: C Cojocariu, M T Hetmanski, P Silcock, R J Fussell
Affiliation: Thermo Fisher Scientific, Food and Environment Research Agency (FERA), York, UK
Uploaded on 6/29/2015.
For Research Use Only. Not for use in diagnostic procedures.