Description
The Thermo Scientific HPLC system is applied for the analysis of gun surveillance standards. The separation is performed on a Thermo Scientific Acclaim Explosives E2 column, with UV detection at 254 nm.| Market: | Environmental |
| Keywords: | Acclaim Explosives E2, Diphenylamine, DPA, Gun surveillance standards, HPLC, UV detection, Gun powder residue |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.