Find methods for your needs
Improved resolution of 18 PAHs including benzo-fluoranthenes by GC-MS
Description
The Thermo Scientific TRACE GC Ultra is applied for the analysis of polycyclic aromatic hydrocarbons (PAHs). The separation was performed on a Thermo Scientific TraceGOLD TG-17SilMS GC column, with MS detection. The unique selectivity of this stationary phase is shown to fully resolve benzo(j)fluoranthene from benzo(b)fluoranthene and benzo(k)fluoranthene.| Market: | Environmental |
| Keywords: | GC, MS detection, Polycyclic aromatic hydrocarbons (PAHs), TraceGOLD TG-17SilMS |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.