Find methods for your needs
Fast analysis of the Florida TRPH C8–C40 standard using conventional GC instrumentation
Description
In this work the performance of a 0.15 mm internal diameter (ID) Thermo Scientific TraceGOLD TG-1MS GC column is compared with that of a 0.25 mm ID GC column. An increase in speed of analysis for a TRPH (Florida) C8–C40 standard is demonstrated with the 0.15 mm column. No compromise in the separation capability of the method was observed and conversion of the conventional method was easily achieved.| Market: | Environmental |
| Keywords: | C8–C40, Fast, FID, Florida TRPH, GC, TraceGOLD TG-1MS |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.