Find methods for your needs
Determination of water soluble vitamins using a HILIC method
Description
The Thermo Scientific Hypersil GOLD HILIC column was used to separate water soluble vitamins using UV detection at 205, 230 and 260 nm in under 8 minutes.| Market: | Food and Beverage |
| Keywords: | 4-Aminobenzoic acid (PABA), HPLC, Hypersil GOLD HILIC, Nicotinamide, Nicotinic acid, Pyridoxine, Riboflavin, Thiamine, UV detection, Water soluble vitamins |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.