Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the analysis of ISO 22478 explosives. The separation is performed on a Thermo Scientific Acclaim Explosives E2 column, with UV detection at 210 and 254 nm.| Market: | Environmental |
| Keywords: | Acclaim Explosives E2, DEGDN, DNB, DNT, DPA, EGDN, Explosives, HMX, HNDPA, HPLC, ISO 22478, NB, NT, PETN, RDX, Tetryl, TNB, TNT, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.