Find methods for your needs
High resolution analysis of explosives for EPA method 8330A
Description
The Thermo Scientific HPLC system is applied for the analysis of explosives by EPA method 8300A. The separation is performed on a Thermo Scientific Acclaim Explosives E2 column, with UV detection at 254 nm.| Market: | Environmental |
| Keywords: | Acclaim Explosives E2, DNB, DNT, EPA 8330A, Explosives, HMX, HPLC, NB, NT, RDX, Tetryl, TNB, TNT, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.