Find methods for your needs
Improved separation of taxanes using a PFP stationary phase HPLC column
Description
The separation of taxanes was performed using a simple reversed-phase gradient method on a Thermo Scientific Hypersil GOLD PFP column with UV detection at 220 nm.| Market: | Pharma |
| Keywords: | HPLC, Hypersil GOLD PFP, Taxanes, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.