Find methods for your needs
Rapid separation of curcuminoids on a Thermo Scientific Accucore Polar Premium column
Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the separation of curcuminoids in turmeric. The separation is performed on a Thermo Scientific Accucore Polar Premium column, with UV detection at 428 nm. The separation achieved is compared to that obtained on a competitor C18 core-shell column.| Market: | Food and Beverage |
| Keywords: | Accucore Polar Premium, Curcumin, Curcuminoids, Desmethoxycurcumin, HPLC, Turmeric, UV detection |
| Matrix: | Turmeric |
For Research Use Only. Not for use in diagnostic procedures.