Find methods for your needs
Improved determination of carotenoid profiles of yam cultivars using a Thermo Scientific Acclaim C30 column
Description
The Thermo Scientific UltiMate 3000 RSLC system is applied for the determination of carotenoid profiles of yam cultivars. The separation is performed on a Thermo Scientific Acclaim C30 column, with UV detection at 450 nm.| Market: | Food and Beverage |
| Keywords: | Acclaim C30, Carotene, Carotenoid, Cryptoxanthin, HPLC, Lutein, Lycopene, UV detection, Yam |
| Matrix: | Yam |
For Research Use Only. Not for use in diagnostic procedures.