Find methods for your needs
Improved analysis of anthocyanins in raspberry using a Thermo Scientific Acclaim C30 column
Description
The Thermo Scientific UltiMate 3000 RSLC system is applied for the analysis of anthocyanins in raspberry. The separation is performed on a Thermo Scientific Acclaim C30 column, with UV detection at 520 nm.| Market: | Food and Beverage |
| Keywords: | Acclaim C30, Anthocyanin, HPLC, Raspberry, UV detection |
| Matrix: | Raspberry |
For Research Use Only. Not for use in diagnostic procedures.