Find methods for your needs
Improved analysis of anthocyanins in blackberry using a Thermo Scientific Acclaim C30 column
Description
The Thermo Scientific UltiMate 3000 RSLC system is applied for the analysis of anthocyanins in blackberry. The separation is performed on a Thermo Scientific Acclaim C30 column, with UV detection at 520 nm.| Market: | Food and Beverage |
| Keywords: | Acclaim C30, Anthocyanin, Blackberry, Cyanidin-3-glucoside, HPLC, UV detection |
| Matrix: | Blackberry |
For Research Use Only. Not for use in diagnostic procedures.