Find methods for your needs
Improved isocratic resolution of antihistamines and their impurities on a Thermo Scientific Acclaim 120 C18 column
Description
The Thermo Scientific HPLC system is applied for the analysis of antihistamines and their impurities. The separation is performed on a Thermo Scientific Acclaim 120 C18 HPLC column, with UV detection at 249 nm.| Market: | Pharma |
| Keywords: | Acclaim 120 C18, Antihistamines, HPLC, Impurities, Phenothiazine, Promethazine, Pyrrobutamine phosphate, Thenyldiamine, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.