Find methods for your needs
Rapid determination of EPA 8330A explosives using a Thermo Scientific Acclaim RSLC Explosives E2 column
Description
The Thermo Scientific UltiMate 3000 RSLC system is applied for the analysis of EPA 8330A explosives. The separation is performed on a Thermo Scientific Acclaim Explosives E2 RSLC column, with UV detection at 254 nm.| Market: | Environmental |
| Keywords: | Acclaim Explosives E2, DNB, DNT, EPA 8330, Explosives, HMX, HPLC, NB, NT, RDX, Tetryl, TNB, TNT, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.