Find methods for your needs
Rapid separation of bisphenol-A diglycidyl ether (BADGE) and related impurities on a Thermo Scientific Acclaim 120 C18 RSLC column
Description
The Thermo Scientific UltiMate 3000 RSLC system is applied for the analysis of bisphenol-A diglycidyl ether (BADGE) and related impurities. The separation is performed on a Thermo Scientific Acclaim 120 C18 RSLC column, with UV detection at 277 and 228 nm.| Market: | Food and Beverage |
| Keywords: | Acclaim 120 C18, BADGE, Bisphenol-A diglycidyl ether, HPLC, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.