Find methods for your needs
High resolution analysis of pyrethoid insecticide using a Thermo Scientific Acclaim 120 C18 RSLC column
Description
The Thermo Scientific UltiMate 3000 RSLC system is applied for the analysis of pyrethoid insecticide. The separation is performed on a Thermo Scientific Acclaim C18 RSLC column, with UV detection at 220 nm.| Market: | Environmental |
| Keywords: | Acclaim 120 C18, Baythroid, Fenvalerate, HPLC, Insecticide, Phenothrin, Pyrethin, Resmethrin, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.