Find methods for your needs
Rapid determination of APIs in Advair using a Thermo Scientific Acclaim RSLC C18 column
Description
The Thermo Scientific UltiMate 3000 RSLC system is applied for the determination of APIs in Advair. The separation is performed on a Thermo Scientific Acclaim C18 RSLC column, with UV detection at 225 nm.| Market: | Pharma |
| Keywords: | Acclaim 120 C18, Advair, APIs, Fluticasone propionate, HPLC, Hydroxynaphthoic acid, Salmeterol |
| Matrix: | Advair |
For Research Use Only. Not for use in diagnostic procedures.