Find methods for your needs
Improved analysis of sudan dyes in paprika using a Thermo Scientific Acclaim 120 C18 HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of sudan dyes in paprika. The separation is performed on a Thermo Scientific Acclaim 120 C18 HPLC column, with UV detection at 500 nm.| Market: | Food and Beverage |
| Keywords: | Acclaim 120 C18, HPLC, Paprika, Sudan dyes, UV detection |
| Matrix: | Paprika |
For Research Use Only. Not for use in diagnostic procedures.