Find methods for your needs
Improved determination of N-methylcarbamates using a Thermo Scientific Acclaim Carbamate HPLC column Be the first to rate this application
Description
The Thermo Scientific UltiMate 3000 RSLC system is applied for the determination of N-methylcarbamates according to EPA Method 531.2. The separation is performed on a Thermo Scientific Acclaim Carbamate HPLC column, with fluorescence detection.
Market: Environmental
Keywords: Acclaim Carbamate, Aldicarb, BDMC, Carbaryl, Carbofuran, EPA method 531.2, Fluorescence detection, HPLC, Hydroxycarbofuran, Methiocarb, Methomyl, Naphthol, Oxamyl, Propoxur
Matrix: N/A
Uploaded on 9/20/2010.
For Research Use Only. Not for use in diagnostic procedures.