Find methods for your needs
Improved analysis of twenty one preservative compounds found in cosmetics using a Thermo Scientific Hypersil GOLD Phenyl column
Description
The Thermo Scientific HPLC system is applied for the analysis of twenty one preservative compounds found in cosmetics. The separation is performed on a Thermo Scientific Hypersil GOLD Phenyl column, with UV detection at 254 and 214 nm.| Market: | Other |
| Keywords: | Cosmetics, HPLC, Hypersil GOLD Phenyl, Parabens, Preservatives, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.