Find methods for your needs
Fast analysis of famotidine and related substances using a Thermo Scientific Acclaim 120 C18 column
Description
The Thermo Scientific HPLC system is applied for the analysis of famotidine and related substances. The separation is performed using a Thermo Scientific Acclaim 120 C18 column, with UV detection at 275 nm.| Market: | Pharma |
| Keywords: | Acclaim 120 C18, Famotidine, HPLC, Impurities, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.