Find methods for your needs
Improved analysis of cetylpyridinium chloride in mouthwash using a Thermo Scientific Acclaim Surfactant column
Description
The Thermo Scientific HPLC system is applied for the analysis of cetylpyridinium chloride in mouthwash. The separation is performed using a Thermo Scientific Acclaim Surfactant column, with suppressed conductivity detection.| Market: | Pharma |
| Keywords: | Acclaim Surfactant, Cetylpyridinium chloride, HPLC, Mouthwash, Suppressed conductivity |
| Matrix: | Mouthwash |
For Research Use Only. Not for use in diagnostic procedures.