 Rate
                        Rate
                     Ask the Expert
                        Ask the Expert
                     Suggest an Application
                            Suggest an Application
                         Add to Favorites
                        Add to Favorites
                     Create
                            CreateParts List
 Tell a
                        Tell a Friend
 Print
                        PrintFind methods for your needs
Description
建立高效的气相色谱质谱检测方法,灵敏、快速测定动 物源性食品中的氯羟吡啶的残留量 样品中的氯羟吡啶用乙腈提取,经碱性氧化铝小柱净化, 衍生后气质联用仪分析 方法检出限远远低于食品安全国家标准中规定的氯羟吡 啶最大残留限量10μg/kg| Market: | Food and Beverage | 
| Keywords: | 单四级杆质谱仪;兽药;碱性氧化铝小柱;TG-5MS 色谱柱 | 
For Research Use Only. Not for use in diagnostic procedures.