Find methods for your needs
Fast analysis of melamine on a Thermo Scientific Acclaim Mixed-Mode WAX-1 HPLC column
Description
The Thermo Scientific Acclaim Mixed-Mode WAX-1 column is uses a combination of HILIC and anion exchange to retain the polar molecule melamine.| Market: | Food and Beverage |
| Keywords: | Acclaim Mixed-Mode WAX-1, HPLC, melamine, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.