Find methods for your needs
A simple determination of Doxorubicin using a Thermo Scientific Accucore C8 Column
Description
The Thermo Scientific Dionex Ultimate 3000 system is applied for the determination of doxorubicin. The separation was performed on a Thermo Scientific Accucore C8 column with UV detection at 254 nm.| Market: | Pharma |
| Keywords: | Accucore C8, Anthracycline, Doxorubicin, HPLC, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.