Find methods for your needs
The reliable extraction and analysis of 18 polycyclic aromatic hydrocarbons in soil using the QuEChERS extraction method and a Thermo Scientific TraceGOLD TG-17SilMS GC column
Description
The Thermo Scientific TRACE GC Ultra is applied to the separation of 18 Polycyclic Aromatic Hydrocarbons using the Thermo Scientific TraceGOLD TG-17SilMS 0.25μm GC column with MS detection| Market: | Environmental |
| Keywords: | benzofluoranthenes b j and k, PAH, Polycyclic Aromatic Hydrocarbon (PAH), QuEChERS, soil, TraceGOLD TG-17SilMS |
| Matrix: | Soil |
For Research Use Only. Not for use in diagnostic procedures.