Find methods for your needs
Analysis of derivatized chlorinated acetic acids using a Thermo Scientific TRACE TR-5 column
Description
The analysis demonstrates the advantages of using the Thermo Scientific TRACE GC Ultra with the Thermo Scientific TRACE TR-5 0.25 μm column for the separation of derivatized chlorinated acetic acids using FID detection.| Market: | Environmental |
| Keywords: | chlorinated acetic acid, chloroacetic acid (CA), Derivatization, dichloroacetic acid (DCA), extractive alkylation, FID, pentafluorobenzyl bromide (PFBBr), trichloroacetic acid (TCA) |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.