Find methods for your needs
Determination of Humulones and Lupulones Be the first to rate this application
Description
The Thermo Scientific Humulones and Lupulones standard was directly injected onto a Thermo Scientific Hypersil GOLD 3 μm column. With a total run time of 20 minutes, the method allows fast screening for remaining humulones (or lupulones). Hence, the isomerization progress during beer brewing can be observed fast and easy.
Market: Food and Beverage
Keywords: Beer, BeerNHop, Bitterness, Brewery, Brewing, Hop, HPLC, HPLC, Isohumulones, Solution
Matrix: Certified humulones, Lupulones standard in methanol (100 µg/mL)
Uploaded on 10/27/2014.
For Research Use Only. Not for use in diagnostic procedures.