Description
The Thermo Scientific Tetrahydroisohumulones standard was directly injected onto a Thermo Scientific Hypersil GOLD 3 μm column. With a total run time of 20 minutes, the method allows fast screening for reduced isohumulones in beer. Hence, beers can be screened if they comply with the German purity law.| Market: | Food and Beverage |
| Matrix: | Certified tetrahydroisohumulones standard in methanol (100 µg/mL) |
For Research Use Only. Not for use in diagnostic procedures.