Find methods for your needs
Fast separation of glyceric, glycolic, lactic, acetic, and levulinic acids on a Thermo Scientific Acclaim Mixed-Mode WAX-1 HPLC column Be the first to rate this application
Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the analysis of glyceric, glycolic, lactic, acetic, and levulinic acids. The separation is performed on a Thermo Scientific Acclaim Mixed-Mode WAX-1 HPLC column in under 7 minutes, with UV detection at 210 nm. This group of very hydrophilic acids are difficult to separate using a reversed-phase column. But on the Thermo Scientific Acclaim Mixed-Mode WAX-1 column, this task becomes easy, as its unique column chemistry combines both reversed-phase and anion-exchange properties on the same column.
Market: Chemical
Keywords: Acclaim Mixed-Mode WAX-1, Acetic acid, Glyceric acid, Glycolic acid, HPLC, Lactic acid, Levulinic acid, UltiMate 3000, UV detection
Matrix: N/A
Uploaded on 2/22/2010.
For Research Use Only. Not for use in diagnostic procedures.