Find methods for your needs
LC-MS/MS method for the rapid determination of Testosterone using a Thermo Scientific Accucore C8 HPLC column
Description
A Thermo Scientific HPLC system coupled with the Thermo Scientific TSQ Vantage mass spectrometer are applied for the rapid determination of testosterone. The separation was performed on a Thermo Scientific Accucore C8 HPLC column, with a cycle time of 1.5 minutes.| Market: | Pharma |
| Keywords: | Accucore C8, LC-MS/MS, Testosterone |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.