Description
本方法采用一种新的柱切换的方式,排除了样品中大浓度的氯离子基体的干扰,对样品中的痕量的溴酸根离子进行了系统的分析测定,6种品牌的饮用水中的溴酸根离子含量都没有超过世界卫生组织颁布的《饮用水质标准标准》中规定的含量。结果表明,所使用的方法真实可靠,重现性良好,分析方法简单,峰形较好,且能与其他离子峰完全分离。| Market: | Water Analysis |
| Keywords: | 离子色谱;电导;溴酸盐;饮用水;阀切换 |
| Matrix: | 饮用水 |
For Research Use Only. Not for use in diagnostic procedures.