Find methods for your needs
毛细管离子色谱法测定饮用水中的常见阴离子 Be the first to rate this application
毛细管离子色谱法测定饮用水中的常见阴离子
Description
采用毛细管离子色谱法测定水中常见阴离子具有灵敏度高、准确性好等优点。进样量仅需0.4 μL,且24小时仅需消耗水14.4 mL,成本很低。
Market: Water Analysis
Keywords: 离子色谱;毛细管离子色谱;阴离子;饮用水
Matrix: 饮用水
Uploaded on 10/18/2014.
For Research Use Only. Not for use in diagnostic procedures.