Description
本文建立了饮用水中痕量消毒副产物的离子色谱测定方法。该法快速简便、分离效果好、灵敏度和准确度高,利用HAAs的阴离子电化学特性达到检测目的,避免了HAAs的衍生化,值得进一步研究和推广使用。该方法可满足饮用水中低含量五种消毒副产物的测定要求。| Market: | Water Analysis |
| Keywords: | 消毒副产物;离子色谱;卤代乙酸 |
| Matrix: | 饮用水 |
For Research Use Only. Not for use in diagnostic procedures.