Description
本文采用吹扫- 捕集- 气质联用法分析水中苯系物、氯苯类化合物及部分卤代烃(苯、甲苯、邻二甲苯、间二甲苯、乙苯、异丙苯、氯苯、1,3- 二氯苯、1,4- 二氯苯、1,2- 二氯苯、1,2,3- 三氯苯、1,3,5- 三氯苯、1,2,4- 三氯苯和1,1,1- 三氯乙烷), 具有简便、快速、灵敏度高、准确性好等特点。满足国标饮用水106 项法规的要求。| Market: | Water Analysis |
| Keywords: | 吹扫捕集;气质联用;饮用水;VOC |
| Matrix: | 饮用水 |
For Research Use Only. Not for use in diagnostic procedures.