Description
采用赛默飞世尔全新一代TRACE 1310 GC 含电子捕获检测器,结合Triplus-RSH 三合一自动进样器 , 本实验使用其中顶空功能。其具有安装快捷方便,同时测定灵敏度高、重复性好、结果可靠等优点,本文完全满足饮用水中多种卤代烃的分析与检测需要,同时可以轻松应对实验室各种对水质分析的要求。| Market: | Water Analysis |
| Keywords: | 顶空法;气相色谱;TG-624 MS;环境水;卤代烃 |
| Matrix: | 环境水 |
For Research Use Only. Not for use in diagnostic procedures.