Description
本实验采用Thermo Scientific ISQ 质谱仪,配合Triplus RSH 自动进样器采用顶空进样,分析烟用纸张中21 种挥发性有机化合物,实验结果完全满足烟草分析方法对杂质限量检测和重现性的要求。| Market: | Other |
| Keywords: | 气相色谱;气质联用仪;挥发性有机物 |
| Matrix: | 烟用纸张 |
For Research Use Only. Not for use in diagnostic procedures.