Description
本方法利用双三元 高效液相系统(DGLC-3600),双三元右泵作为分析泵,而左泵作为衍生泵提供衍生试剂,柱温箱提供衍生反应温度,对黄曲霉毒素柱后衍生后,荧光检测器检测。对5种黄曲霉毒素的检出限可达0.013-0.075 μg/Kg,同时满足标准方法及国标限值的检测要求。| Market: | Food and Beverage |
| Keywords: | 柱后碘衍生;双三元;黄曲霉毒素 |
| Matrix: | 芝麻 |
For Research Use Only. Not for use in diagnostic procedures.