Find methods for your needs
Separation of montelukast and levocetirizine using a reversed-phase HPLC-UV method
Description
The Thermo Scientific Dionex UltiMate 3000 system is applied to demonstrate the separation of montelukast and levocetirizine with a total run time of 22 minutes using the Thermo Scientific Accucore RP-MS 2.6 µm HPLC column.| Market: | Pharma |
| Keywords: | Accucore RP-MS, Antihistamine, HPLC, Levocitrizine, Montelukast, UV detection |
| Matrix: | N/A |
| Author: | Prakash Chander |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.