Find methods for your needs
GCMS法测定白酒中邻苯二甲酸酯类物质残留量 Be the first to rate this application
gcms法测定白酒中邻苯二甲酸酯类物质残留量
Description
本方法采用去除乙醇后正己烷提取,气相色谱质谱联用仪检测的技术,建立了白酒中16种邻苯二甲酸酯类物质残留的分析方法
Market: Food and Beverage
Keywords: GCMS;白酒;邻苯二甲酸酯类物质
Matrix: 白酒
Uploaded on 10/16/2014.
For Research Use Only. Not for use in diagnostic procedures.