Description
离子色谱分离脉冲安培检测器测定果糖二磷酸钠及其有关物质果糖、磷酸葡萄糖和磷酸果糖的方法操作简便,重复性好,线性范围内相关性好,准确度高。同时果糖二磷酸和三种有关物质相比灵敏度较低,在进行含量测定时,无需较大稀释倍数,可以减小由于稀释倍数过大引入的误差。该方法可同时进行含量测定和有关物质分析,具有较高的实用价值。| Market: | Pharma |
| Keywords: | 离子色谱;脉冲积分安培;果糖二磷酸钠 |
| Matrix: | 注射液 |
For Research Use Only. Not for use in diagnostic procedures.