Description
赛默飞世尔科技提供了从样品前处理、仪器分析及报告输出全套解决方案。加速溶剂萃取具有快速,高效,节省溶剂等特点,能够在20min 内将样品萃取完全。2012 年4 月推出的全新一代TRACE 1310 GC 能够完全满足《中国药典(2010 版)》对检测灵敏度,精密度的要求。以及TRACE1310 GC 完全由Chromeleon 7.1 软件控制,该软件直观易懂,简单易用,直接利用自带导航操作即可轻松应对纷繁复杂分析需求。| Market: | Pharma |
| Keywords: | 加速溶剂萃取;固相萃取;气相色谱法;中药;有机氯;农药 |
| Matrix: | 农药 |
For Research Use Only. Not for use in diagnostic procedures.