Description
采用赛默飞世尔加速溶剂萃取和全新一代单四级杆气相色谱质谱联用仪,样品前处理时间极大缩短了,每个样品的提取时间在20min 左右;35min 内能够分析检测49 种农药残留,使得实验室能够应付大批量样品的分析检测| Market: | Pharma |
| Keywords: | 加速溶剂萃取;农药多残留 |
| Matrix: | 农药 |
For Research Use Only. Not for use in diagnostic procedures.