Description
采用ASE提取娑罗子中的七叶皂苷,当采用1次萃取循环作为循环次数时,ASE较2010版药典推荐方法对七叶皂苷的提取率略高;当采用2次萃取循环作为循环次数时,ASE对七叶皂苷的提取效率较药典方法提高了36%。ASE提取中药中的活性成分时,既能达到药典推荐萃取方法等同的萃取率,在提高萃取条件时,ASE还能达到更高的萃取率。同时具有操作简单、快速、高效等特点,值得在中药领域推广使用。| Market: | Pharma |
| Keywords: | 加速溶剂萃取;顺序萃取;中药;娑罗子;七叶皂苷 |
| Matrix: | 中药 |
For Research Use Only. Not for use in diagnostic procedures.