Find methods for your needs
采用加速溶剂萃取(ASE)技术提取黄芪的黄芪甲苷 Be the first to rate this application
采用加速溶剂萃取(ase)技术提取黄芪的黄芪甲苷
Description
ASE是专为需要分析大量样品且从样品中提取有效产品信息的实验室而设计。ASE利用提高温和增加压力可以增加溶剂动力学的原理,萃取效率提高的同时减少溶剂消耗量和萃取时间。在许多应用领域的数据表明,ASE的萃取结果与其它传统萃取方法的结果相似。然而与传统分析相比,ASE萃取效率更高,将原来十几小时的萃取时间缩短至小于1小时甚至更短。
Market: Pharma
Keywords: 加速溶剂萃取;中药;活性成分;黄芪;黄芪甲苷
Matrix: 黄芪
Uploaded on 10/15/2014.
For Research Use Only. Not for use in diagnostic procedures.