Description
本文建立的二维毛细管离子法用于测定有机溶剂中的痕量阴离子,相比于传统的离子色谱法更灵敏、方便和快速,而这些优点主要是因为应用了ICS5000毛细管离子色谱仪器。在前处理部分,基体可以很容易的被去除掉;在分析测试部分,目标阴离子在毛细管系统中分析,可以确保很高的灵敏度,优异的精密度和更短的分析时间。结果显示,本文所建立的系统对各种有机溶剂中痕量阴离子的测定具体很好的应用前景。| Market: | Chemical |
| Keywords: | 毛细管离子色谱;二维色谱分离;有机溶剂;痕量阴离子 |
| Matrix: | 有机溶剂 |
For Research Use Only. Not for use in diagnostic procedures.